Learn More
American Research Products Inc Mouse Factor related Apoptosis
Catalog No. 5000326603
Encompass
Mouse Factor-related Apoptosis FAS ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26603 Supplier American Research Products Inc Supplier No. CSBE04543M
Mouse Factor-related Apoptosis FAS ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich