Learn More
American Research Products Inc Mouse Factor related Apoptosis
Supplier:  American Research Products Inc CSBE04545M
Encompass
Mouse Factor-related Apoptosis ligand FASL ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29895