Learn More
Biomatik Corporation Mouse Family With Sequence Similarity 135, Member B (FAM135B) ELISA Kit
Supplier:  Biomatik Corporation EKU04016
Encompass_Preferred
Mouse Family With Sequence Similarity 135, Member B (FAM135B) ELISA Kit for detecting FAM135B in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9DAI6. Gene ID: 70363. Target Synonyms: C8ORFK32. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5843