Learn More
ACROBiosystems ACROBiosystems Mouse FAP Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems FAPM52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse FAP, His Tag (FAP-M52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asp 761 (Accession # P97321).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 87.2 kDa.
Catalog No. 50-210-6452