Learn More
Cayman Chemical Mouse Farnesoid X Receptor Reporter Assay System, 96 Well
SDP
Catalog No. 502984646
Encompass_Preferred
Mouse FXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. M00601-32, M00601]
Catalog No. 50-298-4646 Supplier Cayman Chemical Supplier No. 2034996 WELL
Mouse FXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. M00601-32, M00601]