Learn More
Sino Biological Recombinant Anti-Fas/CD95/TNFRSF6 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50027R004P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD95 / APO-1 / TNFRSF6 / FAS (rM CD95 / APO-1 / TNFRSF6 / FAS; Catalog#50027-M08H; NP_032013.2; Met1-Arg169) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-1745