Learn More
Sino Biological Anti-Fas/CD95/TNFRSF6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50027RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD95 (rM CD95; Catalog#50027-M08H; NP_032013.2; Met 1-Arg 169). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-4085