Learn More
Sino Biological Anti-Fas Ligand/TNFSF6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101984T1020
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse Fas Ligand / FASLG / CD95L, and purified by antigen affinity chromatography.
Catalog No. 50-241-1412