Learn More
TRIBIOSCIENCE INC Mouse Fast Genotyping System
SDP
Catalog No. NC2591444
Encompass
The Fast Genotyping system is designed for rapid DNA extraction and PCR amplification from mouse tail/toe/ear punch, or other tissue. The PCR product can be directly loaded onto gel for genotype analysis. The kit includes rapid DNA extraction reagent, and PCR mixture.
Catalog No. NC2591444 Supplier TRIBIOSCIENCE INC Supplier No. TBS4033
The Fast Genotyping system is designed for rapid DNA extraction and PCR amplification from mouse tail/toe/ear punch, or other tissue. The PCR product can be directly loaded onto gel for genotype analysis. The kit includes rapid DNA extraction reagent, and PCR mixture.