Learn More
Biomatik Corporation Mouse Fatty Acid Binding Protein 9, Testis (FABP9) ELISA Kit
Supplier:  Biomatik Corporation EKU12122
Encompass_Preferred
Mouse Fatty Acid Binding Protein 9, Testis (FABP9) ELISA Kit for detecting FABP9 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08716. Gene ID: 21884. Target Synonyms: TLBP; TFABP; T-FABP; PERF; T-FABP; PERF15; Testis Lipid Binding Protein; Testis-Type Fatty Acid-Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7188