Learn More
Biomatik Corporation Mouse Fatty Acid Binding Protein 2, Intestinal (FABP2) CLIA Kit
Supplier:  Biomatik Corporation EKU12138
Encompass_Preferred
Mouse Fatty Acid Binding Protein 2, Intestinal (FABP2) CLIA Kit for detecting FABP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 68.6-50,000pg/ml. Sensitivity: 25.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P55050. Gene ID: 14079. Target Synonyms: FABPI; IFABP; I-FABP; Intestinal-type fatty acid-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5334