Learn More
American Research Products Inc Mouse Fatty acid synthase FAS
Catalog No. 5000327562
Encompass
Mouse Fatty acid synthase (FASN) ELISA kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27562 Supplier American Research Products Inc Supplier No. CSBEL008435MO
Mouse Fatty acid synthase (FASN) ELISA kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich