Learn More
Biomatik Corporation Mouse F-Box And Leucine Rich Repeat Protein 3 (FBXL3) ELISA Kit
Supplier:  Biomatik Corporation EKU04059
Encompass_Preferred
Mouse F-Box And Leucine Rich Repeat Protein 3 (FBXL3) ELISA Kit for detecting FBXL3 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8C4V4. Gene ID: 50789. Target Synonyms: FBL3; FBL3A; FBXL3A; F-box/LRR-repeat protein 3A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5981