Learn More
R&D Systems™ Mouse Fc epsilon RI alpha (NP_034314) VersaClone cDNA
Catalog No. RDC0950
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
RDC0950 10 μg
1 options
This item is not returnable. View return policy
Catalog No. RDC0950 Supplier R&D Systems™ Supplier No. RDC0950
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fc receptor, IgE, high affinity I, alpha polypeptide
The Mouse Fc epsilon RI alpha (NP_034314) VersaClone cDNA clone contains the complete ORF for Fc epsilon RI alpha, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Fc epsilon RI alpha insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans