Learn More
Sino Biological Recombinant Mouse Fc epsilon RI Protein (hFc Tag)
Supplier:  Sino Biological 50935M02H1
Encompass_Preferred
A DNA sequence encoding the mouse FCER1A (P20489) (Met1-Gln204) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-3048