Learn More
Sino Biological Recombinant Anti-CD64/FCGR1 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50086R027P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD64 / FCGR1 (rM CD64 / FCGR1; Catalog#50086-M08H; NP_034316.1; Met 1-Pro 297) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-2104