Learn More
Sino Biological Mouse FCGR3 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 50326R01520
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD16 / FCGR3 (rM CD16 / FCGR3; Catalog#50326-M08H; P08508-1; Met 1-Thr 215).
Catalog No. 50-237-3438