Learn More
Sino Biological Recombinant Mouse CD16/FCGR3/Fc gamma RIII Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 50326M27HB20
Encompass_Preferred
A DNA sequence encoding the mouse FCGR3 (P08508.1) (Met1-Thr215) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-3305