Learn More
Sino Biological Mouse FCGR4 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 50036R012C25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD16-2 / FCGR4 (rM CD16-2 / FCGR4; Catalog#50036-M08H; NP_653142.2; Met 1-Gln 203) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-237-3908