Learn More
Sino Biological Recombinant Anti-CD16-2/FCGR4/Fc gamma RIV Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50036R012100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FCGR4 (rM FCGR4; Catalog#50036-M08H; NP_653142.2; Met 1-Gln 203).
Catalog No. 50-255-3532