Learn More
Sino Biological Recombinant Anti-CD16-2/FCGR4/Fc gamma RIV Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 50036R012F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD16-2 / FCGR4 (rM CD16-2 / FCGR4; Catalog#50036-M08H; NP_653142.2; Met 1-Gln 203) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-255-3530