Learn More
Sino Biological Anti-CD16-2/FCGR4/Fc gamma RIV Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50036RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse FCGR4 (rM FCGR4; Catalog#50036-M08H; NP_653142.2; Met 1-Gln 203;). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3528