Learn More
BIOHIPPO Mouse FcgRI Microsample
SDP
Catalog No. 5000315752
Encompass
Mouse FcgRI Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FcgRI in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 249 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse FcgRI Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse FcgRI Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse FcgRI biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15752 Supplier BIOHIPPO Supplier No. BHE15211452
Mouse FcgRI Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FcgRI in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 249 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse FcgRI Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse FcgRI Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse FcgRI biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color