Learn More
Sino Biological Recombinant Mouse FCGRT & B2M Heterodimer Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological CT083M27HB20
Encompass_Preferred
A DNA sequence encoding the mouse FCGRT (NP_034319.2) (Met 1-Ser 297) was fused with a C-terminal polyhistidine tagged AVI tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the mouse B2M (P01887,105Ala/Asp) (Met 1-Met 119) constructed the plasmid 2. The two plasmids were co-expressed and the FCGRT / B2M heterodimer was purified. The purified heterodimer was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-2604