Learn More
Sino Biological Anti-FCRL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50947T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse FCRL1 / FCRH1 (Catalog#50947-M08H; Q8R4Y0-2; Met1-Thr204). FCRL1 / FCRH1 specific IgG was purified by Mouse FCRL1 / FCRH1 affinity chromatography.
Catalog No. 50-255-3045