Learn More
ACROBiosystems ACROBiosystems Mouse FcRn / FCGRT&B2M Heterodimer Protein, His Tag&Twin-Strep Tag (BLI verified)
Catalog No. 502106637
Encompass_Preferred
Recombinant Protein;1MG;Mouse FCGRT&B2M Heterodimer Protein, His Tag&Twin-Strep Tag (BLI verified) (FCM-M52W2) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Ser 297 (FCGRT) & Ile 21 - Met 119 (B2M) (Accession # Q61559-1 (FCGRT) & P01887-1 (B2M)).;Mouse FCGRT&B2M Heterodimer Protein, His Tag&Twin-Strep Tag (BLI verified), produced by co-expression of FCGRT and B2M, has a calculated MW of 32.9 kDa (FCGRT) and 14.7 kDa (B2M). Subunit FCGRT is fused with a polyhistidine tag at the C-terminus and subunit Beta-2 microglobulin (B2M) is fused with Twin-Strep tag at the C-terminus. The reducing (R) protein migrates as 42-48 kDa (FCGRT) and 15-17 kDa (B2M) respectively due to glycosylation.;Immobilized Ipilumimab at 5 μg/mL (100 μL/well) can bind Mouse FCGRT&B2M Heterodimer Protein, His Tag&Twin-Strep Tag (BLI verified) (Cat. No. FCM-M52W2) with a linear range of 0.625-5 μg/mL (QC tested).
Catalog No. 50-210-6637 Supplier ACROBiosystems Supplier No. FCMM52W21MG
Recombinant Protein;1MG;Mouse FCGRT&B2M Heterodimer Protein, His Tag&Twin-Strep Tag (BLI verified) (FCM-M52W2) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Ser 297 (FCGRT) & Ile 21 - Met 119 (B2M) (Accession # Q61559-1 (FCGRT) & P01887-1 (B2M)).;Mouse FCGRT&B2M Heterodimer Protein, His Tag&Twin-Strep Tag (BLI verified), produced by co-expression of FCGRT and B2M, has a calculated MW of 32.9 kDa (FCGRT) and 14.7 kDa (B2M). Subunit FCGRT is fused with a polyhistidine tag at the C-terminus and subunit Beta-2 microglobulin (B2M) is fused with Twin-Strep tag at the C-terminus. The reducing (R) protein migrates as 42-48 kDa (FCGRT) and 15-17 kDa (B2M) respectively due to glycosylation.;Immobilized Ipilumimab at 5 μg/mL (100 μL/well) can bind Mouse FCGRT&B2M Heterodimer Protein, His Tag&Twin-Strep Tag (BLI verified) (Cat. No. FCM-M52W2) with a linear range of 0.625-5 μg/mL (QC tested).