Learn More
Crystal Chem Inc Mouse Ferritin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80636
Encompass
The Mouse Ferritin ELISA is used to quantify levels of mouse ferritin in plasma, serum, and urine using only a 10 µL sample volume. Highlights: Kit uses sample size of only 5 µL. High sensitivity of 2.6 ng/mL. Dynamic range of 12.5 - 400 ng/mL. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7767