Learn More
Abcam Mouse Ferritin ELISA Kit (FTL), 1 x 96 Tests
Supplier:  Abcam AB1577131X96TESTS
Encompass_Preferred
Mouse Ferritin ELISA Kit (FTL) ab157713 is used to quantify Mouse Ferritin with a sensitivity of 64 pg/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0702711