Learn More
American Research Products Inc Mouse Ferritin heaVy chain FT
Catalog No. 5000326963
Encompass
Mouse Ferritin heaVy chain (FTH1) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26963 Supplier American Research Products Inc Supplier No. CSBEL009030MO
Mouse Ferritin heaVy chain (FTH1) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich