Learn More
Immunology Consultants Mouse Fetuin ELISA Kit
SDP
Supplier:  Immunology Consultants E90FET
Encompass_Preferred
The Mouse Fetuin (FET) test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring FET in mouse biological samples.
Catalog No. 50-240-6598