Learn More
Sino Biological Mouse Fetuin A/FETUA Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000785406
Encompass_Preferred
The Mouse Fetuin-A / AHSG ELISA Pair Set is for the quantitative determination of Mouse Fetuin-A / AHSG.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Fetuin-A / AHSG coated on a 96-well plate. Standards and samples are added to the wells, and any Fetuin-A / AHSG present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Fetuin-A / AHSG polyclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Fetuin-A / AHSG present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85406 Supplier Sino Biological Supplier No. SEK5009315
The Mouse Fetuin-A / AHSG ELISA Pair Set is for the quantitative determination of Mouse Fetuin-A / AHSG.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Fetuin-A / AHSG coated on a 96-well plate. Standards and samples are added to the wells, and any Fetuin-A / AHSG present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Fetuin-A / AHSG polyclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Fetuin-A / AHSG present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.