Learn More
Biomatik Corporation Mouse Fetuin B (FETUB) ELISA Kit
Supplier:  Biomatik Corporation EKU11703
Encompass_Preferred
Mouse Fetuin B (FETUB) ELISA Kit for detecting FETUB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.41ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QXC1. Gene ID: 59083. Target Synonyms: FETU-B; 16G2; Gugu; IRL685; Fetuin-like protein IRL685. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7736