Learn More
BIOHIPPO Mouse FFAR4 Microsample
SDP
Catalog No. 5000315657
Encompass
Mouse FFAR4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FFAR4 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 121 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse FFAR4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse FFAR4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse FFAR4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15657 Supplier BIOHIPPO Supplier No. BHE15210973
Mouse FFAR4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FFAR4 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 121 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse FFAR4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse FFAR4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse FFAR4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color