Learn More
BIOHIPPO Mouse FGa Microsample
SDP
Catalog No. 5000315386
Encompass
Mouse FGa Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FGa in Plasma It provides a detection range of 31 25-2000 ng/mL with a sensitivity of 10 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse FGa Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse FGa Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse FGa biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15386 Supplier BIOHIPPO Supplier No. BHE15211304
Mouse FGa Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FGa in Plasma It provides a detection range of 31 25-2000 ng/mL with a sensitivity of 10 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse FGa Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse FGa Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse FGa biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color