Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse FGF-4 Recombinant Protein, PeproTech®

Catalog No. 4505725UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-57-25UG 25 μg
45-057-01M 2 x 500 μg
45057100UG 100 μg
45057250UG 250 μg
45057500UG 500 μg
450-57-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 450-57-25UG Supplier Gibco™ Supplier No. 4505725UG
  • $225.00
    $168.65 / Each of 1

    Save $56.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

450-μ-1MG will be provided as 2 x 500 μg (450-μ-500UG). Recombinant Murine FGF-4 is a 19.0 kDa protein consisting of 173 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.

Fibroblast growth factor 4 (FGF4) is a member of the fibroblast growth factor (FGF) family that possess broad mitogenic and cell survival activities and play key roles in growth and survival of stem cells during embryogenesis, tissue regeneration, and carcinogenesis. FGF4 was identified by its strong oncogenic transforming activity and is a potent angiogenic factor, expressed in several highly vascularized tumors and also in adult mouse testis, intestine, and brain. Studies on the mouse homolog suggests a function in bone morphogenesis and limb development through the sonic hedgehog (SHH) signaling pathway. Furthermore, FGF4 regulates neural progenitor cell proliferation and neuronal differentiation. Recent studies show a growth-promoting role for FGF4 in human embryonic stem cells and a putative feedback inhibition mechanism by a novel FGF4 splice isoform that may serve to promote differentiation at a later stages of development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11403
For Use With (Application) Bioactivity, Control
Formulation Protein with no preservative
Gene ID (Entrez) 14175
Molecular Weight (g/mol) 19 kDa
Name Mouse FGF-4
Purification Method Purified
Quantity 25 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias FGF; FGF4; Fgf-4; Fgfk; Fibroblast growth factor; fibroblast growth factor 4; fibroblast growth factor 4 splice isoform; HBGF-4; heparin secretory transforming protein 1; heparin secretory-transforming protein 1; heparin-binding growth factor 1; heparin-binding growth factor 4; HST; Hst1; hst-1; HSTF1; HSTF-1; human stomach cancer, transforming factor from FGF-related oncogene; kaposi sarcoma oncogene; Kfgf; K-FGF; K-fibroblast growth factor; KS3; oncogene HST; transforming protein KS3
Common Name FGF4
Gene Symbol Fgf4
Biological Activity Determined by the dose-dependent stimulation of the proliferation of Balb/c 3T3 cells. The expected ED50 for this effect is 0.4-1.0 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence APNGTRHAEL GHGWDGLVAR SLARLPVAAQ PPQAAVRSGA GDYLLGLKRL RRLYCNVGIG FHLQVLPDGR IGGVHADTRD SLLELSPVQR GVVSIFGVAS RFFVAMSSRG KLFGVPFFTD ECKFKEILLP NNYNAYESYA YPGMFMALSK NGRTKKGNRV SPTMKVTHFL PRL
Content And Storage -20°C
Activity Determined by the dose-dependent stimulation of the proliferation of Balb/c 3T3 cells. The expected ED50 for this effect is 0.4-1.0 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.