Learn More
FUJIFILM BIOSCIENCES INC Mouse FGF-9, Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Supplier:  FUJIFILM BIOSCIENCES INC 200131MG
Encompass_Preferred
Fibroblast growth factor 9 (FGF-9) is a mitogen and survival factor for nerve and mesenchymal cells. FGF-9 functions as an autocrine and paracrine factor to support the growth and survival of motor neurons and prostate tissue. FGF-9 expression in the gonad is also necessary for sex determination.
Catalog No. 50-990-833
Edge