Learn More
ACROBiosystems ACROBiosystems Mouse FGF R2 (IIIb) protein, His Tag
Supplier:  ACROBiosystems FGBM52H51MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse FGFR2 (IIIb), His Tag (FGB-M52H5) is expressed from human 293 cells (HEK293). It contains AA Pro 39 - Leu 243 (Accession # P21803-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.8 kDa. The protein migrates as 40-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6522