Learn More
Sino Biological Recombinant Mouse basic FGF/FGF2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50037M07E1
Encompass_Preferred
A DNA sequence encoding the mouse Fgf2 (NP_032032.1) (Ala11-Ser154) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-3526