Learn More
Sino Biological Anti-FGF-21 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50421RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse FGF21 (rM FGF21; Catalog#50421-M08H; Q9JJN1; Met1-Ser210). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3275