Learn More
Sino Biological Anti-FGFR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50186T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse FGFR1 / CD331 (rM FGFR1 / CD331; Catalog#50186-M08H; P16092-1; Met1-Glu376). FGFR1 / CD331 specific IgG was purified by Mouse FGFR1 / CD331 affinity chromatography.
Catalog No. 50-241-0891