Learn More
Sino Biological Recombinant Anti-FGFR3 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50071R04620
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FGFR3 / CD333 (rM FGFR3 / CD333; Catalog#50071-M08H; NP_032036.2; Met1-Tyr367).
Catalog No. 50-255-3488