Learn More
Sino Biological Anti-FGFR3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50071T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse FGFR3 / CD333 (Catalog#50071-M08H; NP_032036.2; Met1-Tyr367). FGFR3 / CD333 specific IgG was purified by Mouse FGFR3 / CD333 affinity chromatography.
Catalog No. 50-255-3486