Learn More
Sino Biological Recombinant Mouse CD38 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50191M08H500
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CD38 (NP_031672.2) (Met1-Thr304) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-222-3716