Learn More
Sino Biological Anti-FGFR4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50194RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse FGFR4 / FGF Receptor 4 extracellular domain (rM FGFR4; Catalog#50194-M08H; Met 1-Asp 366; NP_032037.2). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8104