Learn More
Bon Opus Biosciences Recombinant Mouse FGL1 (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP102350UG
Encompass_Preferred
Recombinant Mouse FGL1 (C-mFc) is produced by Human Cells. The target gene encoding Leu23-Ile314 is expressed with a C-mFc tag. Accession# Q71KU9. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1023
Catalog No. 50-217-2625