Learn More
Biomatik Corporation Mouse Fibrillin 1 (FBN1) ELISA Kit
Supplier:  Biomatik Corporation EKU10819
Encompass_Preferred
Mouse Fibrillin 1 (FBN1) ELISA Kit for detecting FBN1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61554. Gene ID: 14118. Target Synonyms: FBN; MASS; MFS1; OCTD; SGS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7319