Learn More
Biomatik Corporation Mouse Fibrinogen Alpha Chain (FGA) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11632
Encompass_Preferred
Mouse Fibrinogen Alpha Chain (FGA) Mini sample ELISA Kit for detecting FGA in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.251ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: E9PV24. Target Synonyms: Fib2; FG-A; FGA; Fibrinopeptide A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6016