Learn More
Biomatik Corporation Mouse Fibrinogen Beta Chain (FGB) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11634
Encompass_Preferred
Mouse Fibrinogen Beta Chain (FGB) Mini sample ELISA Kit for detecting FGB in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000ng/ml. Sensitivity: 7.1ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8K0E8. Gene ID: 110135. Target Synonyms: FG-B; Fibrinopeptide B; Fibrinogen Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8114