Learn More
American Research Products Inc Mouse Fibrinogen Degradation P
Catalog No. 5000328873
Encompass
Mouse Fibrinogen Degradation Product FDP ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-28873 Supplier American Research Products Inc Supplier No. CSBE07941M
Mouse Fibrinogen Degradation Product FDP ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma Method Competitive