Learn More
Crystal Chem Inc Mouse Fibrinogen ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80637
Encompass
The Mouse Fibrinogen ELISA is used to quantify levels of mouse fibrinogen in serum and plasma using only a 3 µL sample volume. Highlights: Kit uses sample size of only 3 µL. High sensitivity of 2.6 ng/mL. Dynamic range of 25 - 800 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7768